Learn More
UVP Blak-Ray™ B-100AP High-Intensity UV Inspection Lamps: Exposure Box
For use with Blak-Ray™ Model B-100AP/R lamp
Marca: UVP 76-0055-01
Dettagli aggiuntivi : Peso : 0.50000kg
Descrizione
- Made of heat resistant plastic
- Cool-Touch™ housing allows users to handle lamp head for a long time
- Features 100 watt longwave (365nm) bulb and ballast base
- At 18 in. (457mm), it produces highly intense center of UV irradiance approximately 5 in. (127mm) in diameter
- Lamp head rotates 360° when attached to base
- Grip handle fits into the transformer base for easy lamp storage and hands-free lamp operation
Specifica
Exposure Box | |
Fluorescing small samples | |
Blak-Ray™ Model B-100AP/R lamp | |
1 Ea. |