Learn More
3M™ V4 Series Replacement Visor
Click to view available options
Da utilizzare con (apparecchiatura):
V4C visor
V4D visor
V4F visor
Lens Material:
Acetate
Polycarbonate
Steel Mesh
Descrizione
- Easy to replace, visor snaps into place
- For use with 3M™ V4D Faceshield
- Replacement clear acetate visor offering good chemical resistant properties
- Acetate visor has good optical properties and an anti-fog coating
- Replacement clear polycarbonate visor offering impact and scratch resistance properties
- Replacement steel mesh visor with a black matt finish providing protection from flying chips and splinters
Specifica
Specifica
Certificazione/Conformità | CE Approved, EN1731 |
Lens Tint | Clear |
Product Type | V4 Series Replacement Visor |
Da utilizzare con (apparecchiatura) | V4C visor |
Lens Material | Steel Mesh |