Learn More
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops
Thermo Scientific™ Vanquish™ Split Sampler Sample Loops come in a variety of sizes to fit your application requirements.
1165.00€
Specifica
Tipo Sample Loop
Visualizza altre specifiche
Prodotti 1
Codice del prodotto Marca Descrizione Versione biocompatibile Da utilizzare con (apparecchiatura) Prezzo Quantità e disponibilità  
Codice del prodotto Marca Descrizione Versione biocompatibile Da utilizzare con (apparecchiatura) Prezzo Quantità e disponibilità  
16371298
Vedere i documenti
Thermo Scientific™
6850.1915
Sample loop, 10 μL, biocompatible Yes Vanquish Split Sampler HT and FT
1165.00€
Descrizione
Automate sample injection into your Thermo Scientific™ Vanquish™ HPLC or UHPLC system with Thermo Scientific™ Vanquish™ Split Sampler Sample Loops. These sample loops are available in 10, 25, 100, 250, 1000 μL sizes to meet your application requirements. Thermo Scientific™ Viper™ Fittings are included with our sample loops to make them easy to install with leak-free connections.
Vanquish Split Sampler Sample Loops use Viper Fittings, which have a ferrule-free tightening design, for virtually zero-dead-volume connections.
Specifica
Specifica
Sample Loop