Learn More
Emma Safety Footwear Vera Safety Shoes
An easy shoe for wipe clean hygiene. The upper is completely washable microfiber construction. Sanitized with Hydro-Tec™ Sanitized Silver. Stain, fat, grease and direct resistant.
Marca: Emma Safety Footwear MM50654439
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- S2-Safety shoe for wet working conditions
- GripForce™ Easy Twist
- Microfiber construction
- ESD certified
- Chrome-free lining
- Hydro-Tec™ Sanitized Silver
Specifica
EN ISO 20345:2011 | |
White | |
Hydro-Tec™ Sanitized Silver | |
39 |
Elastic | |
Vera | |
Microfiber | |
Steel |