Learn More
3M™ Versaflo™ Filter Cover TR-6100FC
3M™ Versaflo™ Filter Cover TR-6100FC for use with TR-6110E and TR-6130E filters.
Marca: 3M™ 7100021248
12.55 EUR valido fino al 2024-12-31
Usa il codice promo "22912" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 1.00000kg
Descrizione
- Filter cover for use with the TR-6110E and TR-6130E filters
- Filter cover helps protect the filters from physical damage
- Enables use of prefilters and/or spark arrestors
- Enables turbo to be worn through decontamination showers
Specifica
CE Approved | |
TR-6110E and TR-6130E filters | |
0.26 x 0.127 x 0.133 m |
Filter Cover | |
TR-6100FC |