Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Polarizer Set Only for VisiLED Ring Lights
The VisiLED polarizer set is specially designed only for use with the VisiLED brightfield ring light. Due to its cutting edge polarization filters, this set provides a large improvement compared to other polarization tools.
Marca: SCHOTT AG Lighting and Imaging™
Dettagli aggiuntivi : Peso : 28.00000kg
Descrizione
VisiLED Polarization system designed specifically for use only with the VisiLED brightfield ring light (S80-65N-2).
- Turnable polarizer with adapting thread and sliding filter as an analyzer
- Polarized light by a polarizing and analyzing filter within the beam path can be used efficiently in stereomicroscopy to contrast inner structure of material or to identify metal particles
- Not for use with any other ring light
Specifica
Filter | |
A Wide Variety of Tasks From Life Science to Industrial Applications | |
VisiLED Ringlight S80-65N-2 | |
Polarizer Filter and Analyzer |