Learn More
SCHOTT AG Lighting and Imaging™ VisiLED Segmented Polarizer Set for Ring Light S80-55
The VisLED Segmented Polarizer Set has the ability to switch between polarized and non-polarized light, without any mechanical movements.
Marca: SCHOTT AG Lighting and Imaging™ 400555
Dettagli aggiuntivi : Peso : 0.10540kg
Descrizione
- Polarization system for VisiLED Brightfield Ring Light S80-55.
- Switch between polarized and non-polarized light without any mechanical movements.
- Very helpful for all applications that need polarized and non-polarized light (e.g. cleanliness inspection).
- Software control possible with controller MC1500.
- Intensities independently adjustable for polarization and brightfield with controller MC1500.
- Use only with VisiLED Ring Light S80-55 and with controller MC1100 or MC1500 (need to be ordered separately).
Specifica
Polarization Filter | |
A Wide Variety of Tasks From Life Science to Industrial Applications | |
Polarizer Filter and Analyzer |
Polarizer M110 x 1 Analyzer for 66 mm lens, with 4 segments with polarization and 4 segments without polarization | |
VisiLED Ringlight S80-55 | |
M110 x 1 |