missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VUF 8430 dihydrobromide, Tocris Bioscience™
Click to view available options
Quantità:
10 mg
50 mg
Descrizione
High affinity (pKi = 7.5), potent histamine H4 receptor full agonist (pEC50 = 7.3). Displays moderate affinity for H3 receptors (pKi = 6.0) and weak partial agonist activity at H2 receptors.
Specifica
Specifica
Materiale o nome chimico | VUF 8430 dihydrobromide |
Quantità | 50 mg |
Peso formulazione | 323.05 |
Percent Purity | >98% |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'