missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WBP1L Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA558038
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Immunogen sequence: PLSEPSRSST RPPSIADPDP SDLPVDRAAT KAPGMEPSGS VAGLGELDPG AFLDKDAECR EELLKDDSSE HGAPDSKEKT PGRHRRFTGD Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 76%.
WBP1L (OPAL1, outcome predictor in acute leukemia 1) is an almost uncharacterized transmembrane adaptor protein expressed mainly by megakaryocytes and platelets. Its expression is enhanced on TEL/AML leukemic cells. The function of OPAL1 is unknown, although the presence of a cytochrome c-like heme-binding site and a transmembrane domain suggested OPAL1 may be involved in the mitochondrial electron transport chain. Its originally reported prognostic impact appears to be treatment dependent.Specifica
WBP1L | |
Polyclonal | |
Unconjugated | |
WBP1L | |
C10orf26; OPA1L; OPAL1; Outcome predictor in acute leukemia 1; WBP1L; WW domain binding protein 1-like | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54838 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NX94 | |
WBP1L | |
Recombinant protein corresponding to Human WBP1L | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only