Learn More
Cytiva Whatman™ Roby 25 Syringe Filter Validation Kit
For use with Whatman Roby 25 Syringe Filters for Automation (cat. no. 09302128)
Marca: Cytiva 10463898
Dettagli aggiuntivi : Peso : 1.02000kg
Descrizione
- Includes six different types of filters: packed in six tubes with 25 filter holders each
- Filter validation protocol with filter selection aid
- All important selection tests explained step-by-step with all the important properties at a glance
Fine filtration of samples in the automatic tablet dissolution test. The Roby 25 Filter Validation Kit can be used for method development.
Specifica
For automation | |
GF (1μm), GF (0.7μm); 0.45μm | |
150/Pk. | |
Filter Validation Kit |