Learn More
DWK Life Sciences Wheaton™ Borosilicate Glass Coliwasa Samplers PTFE Seals
Used to collect composite liquid samples from top to bottom in drums, tanks and reservoirs
Marca: DWK Life Sciences 885232
57.90 EUR valido fino al 2024-12-31
Usa il codice promo "23261" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 0.00910kg
Descrizione
Positive PTFE seal for borosilicate glass coliwasa.
Specifica
42 in. | |
Seal | |
For Borosilicate Glass coliwasa |
Used to collect composite liquid samples from top to bottom in drums, tanks and reservoirs | |
PTFE | |
106.5 cm |