Learn More
Schmidlin™ Whisper-0 Nitrogen Generator
This nitrogen generator produces a continuous supply of high purity nitrogen.
Marca: Schmidlin™ DB-WN2-0-40
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Increase efficiency and improve safety and performance
- On-demand supply 24/7
- Flow rate at 8 barg (116 psig)
- Purity: LC/MS grade
- Proprietary membrane technology
- No noise, no moving parts and no electricity
- Ideal for all LC/MS applications
Specifica
30 cm | |
130 cm | |
7 bar | |
48 cm | |
>97% at 40 L/min., >99% at 20 L/min. |
40 mL/min. | |
35°C | |
18.9 in. | |
Inlet: 3/8 in., Outlet: 1/4 in. |