Learn More
Buerkle™ White Disposable Forceps
Sfoglia Tutto Bürkle Prodotti
Click to view available options
Stile punta:
punta
punta larga
punta/angolo
Sterilità:
Non-sterile
Sterile
Dimensione della confezione:
pezzi 100
Descrizione
- The fluted handles ensure optimum and secure handling as well as precise work
- The light plastic forceps for single use allow for fatigue-free work
- The clean room-manufactured lab assistants meet the highest standards and prevent cross-contamination or impurities in the sample being examined
- The lab tweezers are suitable for chemical, pharmaceutical or cosmetic applications as well as the food industry and scientific uses
- Sterile forceps are individually wrapped
- For single use
- Cleanroom manufactured (clean room class 7)
- Complies with EU food requirements and FDA regulations
- Pointed/angled tip: The disposable forceps have fine, smooth, curved gripping tips. Different materials can be held and processed reliably with the pointed ends
- Pointed tip: The anatomical forceps have a straight tip, fluted on the inside. This makes it possible to grip sensitive materials reliably
- Broad tip: The disposable forceps have ends with a broad tip. The broad tip is smooth and blunt. It prevents the material from being damaged when it is gripped
Specifica
Specifica
| Stile punta | punta larga |
| N. per confezione | 100 (large pack) |
| Lunghezza (sistema imperiale) | 5.12 in. |
| Materiale | Polystyrene |
| Lunghezza (sistema metrico) | 130 mm |
| Product Type | Pinza |
| Sterilità | Non-sterile |
| Stile | Standard |