missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ White Plastic Bio-Hazard Trays
Available in general purpose, biohazard and radiation safety formats, spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection.
Marca: Fisherbrand™ CSR-TO5
Descrizione
- Available in general purpose, biohazard and radiation safety formats
- Spilltrays provide a re-usable work area with the added benefit of safe containment of spillages and lab bench protection
- All trays supplied with one free liner when purchased
- Additional environmentally friendly APET liners are provided in packs of 25
Specifica
70 x 46 cm | |
70 cm | |
Spill Containment Tray | |
Safe containment of spillages, lab bench protection | |
1 Each |
White | |
Plastic | |
46 cm | |
Liner | |
Rectangular |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto