Learn More
3M™ White Polypropylene Fabric Protective Coverall
Click to view available options
Size: XX-Large
Specifica
Specifica
Product Type | General Purpose Coveralls |
Sesso | Unisex |
Monouso | Single-use |
Stile vita | Elastic |
Ankle Style | Elastic |
Stile polso | Elastic |
Stile collo | Hooded |
Hood Type | 2-panel |
Tipo di giuntura | Serged |
Sterilità | Non-sterile |