Learn More
Smeg™ WP300 Resin Column Water Purifier
Click to view available options
:
WP300 Resin Column Water Purifier
Descrizione
- System uses mixed bed resins (disposable) that can also retain silica
- When resins run out, they can simply be replaced with MI700WP kit
- Resin cartridge is made out of stainless steel, as is entire machine
Specifica
Specifica
| Product Type | WP300 Resin Column Water Purifier |