Learn More
ABEBA™ X-Light Black/Orange Safety Trainers S1 ESD
Black/orange suede X-light safety trainer with silver ion lining to reduce odor, replaceable active comfort insole, steel toe cap, PU slip resistant outsole
Marca: ABEBA™ 7131050/40
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Lightweight, comfortable black/orange trainers
- Comply to EN ISO 20345: 2012, S1 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant Mono PU outsole SRC
- Steel toe cap (200 J)
- Black and orange suede microfiber
- Silver ions lining
- Replaceable active comfort insole
Specifica
Black, Orange | |
40 | |
Comply to EN 20345: 2012 S1 SRC, ESD dissipative according EN 61340 | |
Trainers | |
S1 SRC ESD |
Suede Microfiber | |
Steel | |
Unisex | |
No |