Learn More
Thermo Scientific™ Yersinia Selective Agar
Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples on this prepared media plate after 18 to 24 hours incubation at 32°C.
Marca: Thermo Scientific™ PO5044A
Dettagli aggiuntivi : Peso : 0.30000kg
Descrizione
Isolate and enumerate Yersinia enterocolitica from clinical specimens and food samples after 18 to 24 hours using Thermo Scientific™ Yersinia Selective Agar (CIN Agar).
Specifically developed for the optimum growth and recovery of Yersinia enterocolitica, the formulation includes cefsulodin, Irgasan™ and novobiocin. The typical colonies of Yersinia enterocolitica will develop as a red bullseye surrounded by a transparent border and will vary considerably among serotypes in colony size, smoothness and the ratio of the border to center diameter. Most other organisms that are capable of growing will produce larger colonies (>2mm in diameter) with diffuse pinkish centers and opaque outer zones. Serratia liquefaciens, Citrobacter freundii and Enterobacter agglomerans may give a colonial morphology resembling Yersinia enterocolitica. These organisms can be differentiated from Yersinia enterocolitica by biochemical tests.Our products are manufactured and distributed across the world through a vast network of company-owned facilities to ensure you receive the freshest, highest performing medium possible. For fastest delivery we recommend you select the product part number with a manufacturing origin closest to you.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
Specifica
Tested in accordance with CEN ISO/TS 11133-2.The formulation of this medium conforms to ISO 10273. | |
2° to 12°C, in the dark | |
90mm Monoplate | |
Agar | |
Flow Wrapped - Single Layer |
Antique pink, transparent | |
Yersinia Selective Agar | |
Germany | |
17g ± 5% | |
10/Pk. |