Learn More
ZEOsphere™
Click to view available options
Dimensione del poro:
100 Å
120 Å
300 Å
Fase:
C18 Functionalized
C4 Functionalized
C8 Functionalized
DIOL Functionalized
NH2 Functionalized
Quantità:
100 g
500 g
1 kg
Dimensione della confezione:
100g
1kg
500g
Descrizione
- High performance Spherical Silica Gel
- High Purity
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- Wide Selection of functionalization options
- High Chemical and mechanical stability
- 100 A, 120 A, and 300 A pore sizes available
- 10 μm particle size
- Suitable for medium to high pressure
- High loadability
- Unsurpassed Service and Support
- Selection of pack sizes available
- Suitable for gravity, flash and process
Specifica
Specifica
| Dimensione particelle | 10 μm |
| Dimensione del poro | 100 Å |
| Tecnica | Chromatography |
| Quantità | 1 kg |
| Tipo | Silica Gel |
| Materiale | Gel di silice |
| Fase | C8 Functionalized |
| Forma particelle | Spherical |