missing translation for 'onlineSavingsMsg'
Learn More

Zinc acetate dihydrate, ACS, 98.0-101.0%, Thermo Scientific Chemicals

49.65€ - 290.45€

Identificatori chimici

CAS 5970-45-6
Formula molecolare C4H10O6Zn
Molecular Weight (g/mol) 219.498
Numero MDL MFCD00066961
InChI Key BEAZKUGSCHFXIQ-UHFFFAOYSA-L
Sinonimo zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
PubChem CID 2724192
IUPAC Name zinc;diacetate;dihydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
Visualizza altre specifiche

Prodotti 3
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
11357277
Vedere i documenti
Thermo Scientific Alfa Aesar
011559.30
250 g
49.65€
250g
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
11367277
Vedere i documenti
Thermo Scientific Alfa Aesar
011559.A1
1 kg
86.43€
1kg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
11375889
Vedere i documenti
Thermo Scientific Alfa Aesar
011559.A7
5 kg
290.45€
5kg
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

Zinc acetate dihydrate is used in the preparation of layered Zn-arylphosphonates with potential application in sorption, ion exchange or catalysis and zinc sulfide nanoparticles coated on silica particles. It is used in dietary and medical applications. It is also used as lozenges for treating the common cold. It is used as a seeding material to grow zinc oxide nanorods on the microfibers of polyethylene terephthalate fabric. In addition, it finds application as wood preservation, production of other zinc salts, ethylene acetate manufacture, and an analytical reagent in various industries. As a plating inhibitor, it is used in nuclear power plant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifica

Identificatori chimici

5970-45-6
219.498
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
2724192
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
C4H10O6Zn
MFCD00066961
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
zinc;diacetate;dihydrate

Specifica

Zinc Acetate Dihydrate
5970-45-6
98 to 101.0%
MFCD00066961
zinc acetate dihydrate, zinc diacetate dihydrate, zinc ii acetate dihydrate, unii-fm5526k07a, acetic acid zinc salt, zinc acetate usp, acetic acid, zinc salt, dihydrate, zinc acetate, dihydrate, galzin tn
BEAZKUGSCHFXIQ-UHFFFAOYSA-L
zinc;diacetate;dihydrate
2724192
ACS
237°C
Crystalline
C4H10O6Zn
3732513
Soluble in water,alcoholSoluble in water and alcohol.
CC(=O)[O-].CC(=O)[O-].O.O.[Zn+2]
219.498
219.50 (183.47 Anhydrous)
1.735
Videos
Sicurezza e movimentazione

Sicurezza e movimentazione

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing. This material and/or its container must be disposed of as hazardous waste. Avoid release to the environment. Refer to special instructions/Safety data.

P264b-P270-P280i-P301+P312-P305+P351+P338-P330-P501c

H302-H319

missing translation for 'dotInformation' : DOT Class: 9; Packaging Group: III

missing translation for 'einecsNumber' : 209-170-2

missing translation for 'rtecsNumber' : ZG8750000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Avvertenza
  • Pericolo

Categoria di pericolo
  • Tossicità acuta Categoria 4
  • PERICOLO A LUNGO TERMINE PER L'AMBIENTE ACQUATICO Cronico 2
  • Lesione oculare grave/irritazione oculare Categoria 1

Indicazioni di pericolo
  • H302-Nocivo se ingerito.
  • H318-Provoca gravi lesioni oculari.
  • H411-Tossico per gli organismi acquatici con effetti di lunga durata.

Consigli di prudenza
  • P280-Indossare guanti/indumenti protettivi/Proteggere gli occhi/il viso.
  • P301+P330+P331-IN CASO DI INGESTIONE: sciacquare la bocca. NON provocare il vomito.
  • P305+P351+P338-IN CASO DI CONTATTO CON GLI OCCHI: sciacquare accuratamente per parecchi minuti. Togliere le eventuali lenti a contatto se è agevole farlo. Continuare a sciacquare.
  • P310-Contattare immediatamente un CENTRO ANTIVELENI/un medico .

SDS
Documenti

Documenti

Product Certifications

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato