Learn More
Thermo Scientific Chemicals Zirconium Cover for Crucible
Codice prodotto. 15451397
Click to view available options
Capacità (sistema metrico):
5 mL
10 mL
15 mL
20 mL
25 mL
35 mL
45 mL
55 mL
100 mL
250 mL
500 mL
1000 mL
Descrizione:
For 35884 crucible
For 35888 crucible
For 35905 crucible
For 35909 crucible
For 35926 crucible
For 35930 crucible
For 35947 crucible
For 35951 crucible
For 35968 crucible
For 35972 crucible
For 35997 crucible
For 36014 crucible
For 36035 crucible
For 36039 crucible
For 36054 crucible
For 36058 crucible
For 36070 crucible
For 36083 crucible
For 36087 crucible
For 36097 crucible
Unit Size:
Pezzo
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15451397
Brand: Thermo Scientific Chemicals 036001.KT
Richiesta di formato massivo o personalizzato
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
View alternative products
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Descrizione For 35884 crucible
Materiale Zirconium
Capacità (sistema metrico) 15 mL
Altezza (sistema metrico) Depth:34mm
Product Type Zirconium Crucible Cover