Learn More
Cytiva (formerly Pall Lab) AcroPak™ 500 Capsules with Supor™ Membrane, Sterile
AcroPak capsules designed for research applications only.
Marca: Cytiva (formerly Pall Lab) 12993
Dettagli aggiuntivi : Peso : 0.13600kg
Descrizione
- For buffers, tissue culture media, and other biological fluids
- Newer technology recommended for simplified scale-up
- High filtration area and compact size are ideal for upscale trials
Specifica
Hydrophilic PES | |
Sterile | |
1/Ea. |
0.8/0.45 μm | |
500 cm2 | |
Stepped Hose Barb |