CAS RN 10025-69-1
CAS RN 10025-69-1
Tin(II) chloride dihydrate, 98+%, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 Numero MDL: MFCD00149863 Sinonimo: Stannous chloride dihydrate
Promozioni disponibili
Tin(II) chloride dihydrate, 98%, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Numero MDL: MFCD00149863 Sinonimo: Stannous chloride dihydrate
Tin(II) chloride dihydrate, 98+%, ACS reagent, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 Numero MDL: MFCD00149863 Sinonimo: Stannous chloride dihydrate
Tin(II) Chloride Dihydrate, Certified AR for Analysis, Fisher Chemical™
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Numero MDL: 149863
Promozioni disponibili
Tin(II) chloride dihydrate, low in Mercury, suitable for AAS, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 Sinonimo: Stannous chloride dihydrate
Tin(II) chloride dihydrate, 97%, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Molecular Weight (g/mol): 225.63 Numero MDL: MFCD00149863 Sinonimo: Stannous chloride dihydrate
Tin(II) chloride dihydrate, ACS, 98.0-103.0%, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: SnCl2·2H2O Numero MDL: MFCD00149863
Tin(II) chloride dihydrate, Reagent Grade, Thermo Scientific Chemicals
CAS: 10025-69-1 Formula molecolare: SnCl2·2H2O Numero MDL: MFCD00149863
Tin(Ii) Chloride Dihydrate, puriss. p.a. ACS Reagent, Honeywell Fluka™
CAS: 10025-69-1 Numero MDL: MFCD00149863
Tin(II) Chloride Dihydrate, 98%, ACS Reagent, Honeywell Fluka™
CAS: 10025-69-1 Numero MDL: MFCD00149863
Tin(II) Chloride Dihydrate, Extra Pure, SLR, Fisher Chemical™
CAS: 10025-69-1 Formula molecolare: Cl2Sn·2H2O Numero MDL: 149863
Stannous Chloride, Dihydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
CAS: 10025-69-1 Formula molecolare: SnCl2·2H2O
Tin(Ii) Chloride Dihydrate, Reagent grade, Honeywell™
CAS: 10025-69-1 Numero MDL: MFCD00149863