CAS RN 1005201-24-0
CAS RN 1005201-24-0
IUPAC Name:
2-[3-(4-methanesulfinylphenyl)-1-benzofuran-5-yl]-5-methyl-1,3,4-oxadiazole
Sinonimi:
2-[3-(4-methanesulfinylphenyl)-1-benzofuran-5-yl]-5-methyl-1,3,4-oxadiazole
Peso molecolare (g/mol):
338.38
Formula molecolare:
C18H14N2O3S
InChi Key:
RCKYSTKYIVULEK-UHFFFAOYNA-N
SMILES:
CC1=NN=C(O1)C1=CC=C2OC=C(C2=C1)C1=CC=C(C=C1)S(C)=O
1
–
1
di
1
risultati
1
TCS 2002, Tocris Bioscience™
CAS: 1005201-24-0 Formula molecolare: C18H14N2O3S Molecular Weight (g/mol): 338.381 InChI Key: RCKYSTKYIVULEK-UHFFFAOYSA-N Sinonimo: 2-3-4-methanesulfinylphenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole,mmbo,1,3,4-oxadiazole, 2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl,1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzo b furanyl,2-3-4-methanesulfinyl phenyl-1-benzofuran-5-yl-5-methyl-1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-1-benzofuran-5-yl-1,3,4-oxadiazole,2-methyl-5-3-4-methylsulfinyl phenyl-5-benzofuranyl-1,3,4-oxadiazole PubChem CID: 24855958 IUPAC Name: 2-methyl-5-[3-(4-methylsulfinylphenyl)-1-benzofuran-5-yl]-1,3,4-oxadiazole SMILES: CC1=NN=C(O1)C2=CC3=C(C=C2)OC=C3C4=CC=C(C=C4)S(=O)C