CAS RN 10130-87-7
CAS RN 10130-87-7
IUPAC Name:
2-methoxybenzene-1-sulfonyl chloride
Sinonimi:
2-methoxybenzenesulfonyl chloride
Peso molecolare (g/mol):
206.64
Formula molecolare:
C7H7ClO3S
InChi Key:
GYOBZOBUOMDRRN-UHFFFAOYSA-N
SMILES:
COC1=CC=CC=C1S(Cl)(=O)=O
1
–
1
di
1
risultati
1
2-Methoxybenzenesulfonyl chloride, 95%, Thermo Scientific Chemicals
CAS: 10130-87-7 Formula molecolare: C7H7ClO3S Molecular Weight (g/mol): 206.64 Numero MDL: MFCD01961367 InChI Key: GYOBZOBUOMDRRN-UHFFFAOYSA-N Sinonimo: 2-methoxy-benzenesulfonyl chloride,2-methoxybenzene-1-sulfonyl chloride,2-methoxybenzenesulfonylchloride,o-anisolesulfonyl chloride,2-methoxyphenylsulfonyl chloride,acmc-1bntp,methoxyphenylsulfonyl chloride,2-methoxybenzensulfonylchloride,2-methoxybenzenesulphonylchloride,2-methoxybenzene sulfonylchloride PubChem CID: 3745851 IUPAC Name: 2-methoxybenzenesulfonyl chloride SMILES: COC1=CC=CC=C1S(=O)(=O)Cl