CAS RN 1013937-63-7
CAS RN 1013937-63-7
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
2
di
2
risultati
1
Tocris Bioscience™ VTP 27999 trifluoroacetate
Highly potent and selective renin inhibitor
2