CAS RN 101975-10-4
CAS RN 101975-10-4
IUPAC Name:
6-[4-(difluoromethoxy)-3-methoxyphenyl]-2,3-dihydropyridazin-3-one
Sinonimi:
zardaverine zardaverina zardaverinum
Peso molecolare (g/mol):
268.22
Formula molecolare:
C12H10F2N2O3
InChi Key:
HJMQDJPMQIHLPB-UHFFFAOYSA-N
SMILES:
COC1=CC(=CC=C1OC(F)F)C1=NNC(=O)C=C1
1
–
1
di
1
risultati
1
Zardaverine, Tocris Bioscience™
CAS: 101975-10-4 Formula molecolare: C12H10F2N2O3 Molecular Weight (g/mol): 268.22 InChI Key: HJMQDJPMQIHLPB-UHFFFAOYSA-N Sinonimo: zardaverine,zardaverina,zardaverinum,zardaverine inn,zardaverinum inn-latin,unii-tq358gwh6y,zardaverina inn-spanish,6-4-difluoromethoxy-3-methoxy-phenyl-2h-pyridazin-3-one,6-4-difluoromethoxy-3-methoxyphenyl-3 2h-pyridazinone,tq358gwh6y PubChem CID: 5723 ChEBI: CHEBI:46548 IUPAC Name: 3-[4-(difluoromethoxy)-3-methoxyphenyl]-1H-pyridazin-6-one SMILES: COC1=C(C=CC(=C1)C2=NNC(=O)C=C2)OC(F)F