CAS RN 1020315-31-4
CAS RN 1020315-31-4
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
—
Formula molecolare:
—
InChi Key:
—
SMILES:
—
1
–
4
di
4
risultati
1
PF 04457845
Potent and selective irreversible FAAH inhibitor
2
PF-04457845, MedChemExpress
MedChemExpress PF-04457845 is a highly efficacious and selective FAAH inhibitor with IC50 values is 7.2±0.63 nM and 7.4±0.62 nM for hFAAH and rFAAH, respectively.
3
4