CAS RN 10210-68-1
CAS RN 10210-68-1
IUPAC Name:
—
Sinonimi:
—
Peso molecolare (g/mol):
341.95
Formula molecolare:
C8Co2O8
InChi Key:
ZUDHYLQHRXCZGU-UHFFFAOYSA-N
SMILES:
[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].O=[C-]1[Co][C-](=O)[Co++]1
1
–
2
di
2
risultati
1
Dicobalt octacarbonyl, 95%, stabilized, Thermo Scientific Chemicals
CAS: 10210-68-1 Formula molecolare: C8Co2O8 Molecular Weight (g/mol): 341.95 Numero MDL: MFCD00016024 Sinonimo: carbon monoxide; cobalt 2+,dicobalt octacarbonyl stabilised with hexane,cobalt carbonyl, moistened with hexane hexane co
2
Octacarbonyldicobalt, stab. with 1-5% hexane, Thermo Scientific Chemicals
CAS: 10210-68-1 Numero MDL: MFCD00016024 Sinonimo: carbon monoxide; cobalt 2+,dicobalt octacarbonyl stabilised with hexane,cobalt carbonyl, moistened with hexane hexane co