CAS RN 103-82-2
CAS RN 103-82-2
IUPAC Name:
2-phenylacetic acid
Sinonimi:
ω-phenylacetic acid ω-phenylacetate α-toluic acid phenyllacetic acid α-toluate kyselina fenyloctova w-phenylacetic acid benzeneacetiic acid phenylacetic acid (natural) w-phenylacetate
Peso molecolare (g/mol):
136.15
Formula molecolare:
C8H8O2
InChi Key:
WLJVXDMOQOGPHL-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC=CC=C1
1
–
1
di
1
risultati
1
Phenylacetic acid, 98.5%, Thermo Scientific Chemicals
CAS: 103-82-2 Formula molecolare: C8H8O2 Molecular Weight (g/mol): 136.15 InChI Key: WLJVXDMOQOGPHL-UHFFFAOYSA-N IUPAC Name: 2-phenylacetic acid SMILES: OC(=O)CC1=CC=CC=C1