CAS RN 10361-44-1
CAS RN 10361-44-1
IUPAC Name:
bismuth(3+) trinitrate
Sinonimi:
bismuth(3+) trinitrate
Peso molecolare (g/mol):
394.99
Formula molecolare:
BiN3O9
InChi Key:
PPNKDDZCLDMRHS-UHFFFAOYSA-N
SMILES:
[Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
1
–
1
di
1
risultati
1
Bismuth(III) nitrate hydrate, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
CAS: 10361-44-1 Formula molecolare: BiN3O9 Molecular Weight (g/mol): 394.99 Numero MDL: MFCD00003460 InChI Key: PPNKDDZCLDMRHS-UHFFFAOYSA-N Sinonimo: bismuth nitrate,bismuth trinitrate,ccris 7110,nitric acid, bismuth 3+ salt,tris nitrooxy bismuthine,bismuth 3+ trinitrate,bi.3no3,bismuth iii nitrate,bismuth iii nitrate hydrate,nitric acid, bismuth 3+ salt 3:1 PubChem CID: 107711 IUPAC Name: bismuth;trinitrate SMILES: [Bi+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O