CAS RN 103669-00-7
CAS RN 103669-00-7
IUPAC Name:
[3-(thiophen-2-yl)phenyl]methanol
Sinonimi:
[3-(thiophen-2-yl)phenyl]methanol
Peso molecolare (g/mol):
190.26
Formula molecolare:
C11H10OS
InChi Key:
SATFADRDDGOLJP-UHFFFAOYSA-N
SMILES:
OCC1=CC=CC(=C1)C1=CC=CS1
1
–
1
di
1
risultati
1
(3-Thien-2-ylphenyl)methanol, ≥97%, Thermo Scientific™
CAS: 103669-00-7 Formula molecolare: C11H10OS Molecular Weight (g/mol): 190.26 Numero MDL: MFCD06203108 InChI Key: SATFADRDDGOLJP-UHFFFAOYSA-N Sinonimo: 3-thien-2-ylphenyl methanol,3-2-thienyl benzyl alcohol,3-thiophen-2-yl phenyl methanol,3-2-thienyl phenyl methanol,benzenemethanol,3-2-thienyl,acmc-1c9kv,3-thiophen-2-ylphenyl methanol,3-thiophen-2-yl benzyl alcohol,3-2-thienyl phenyl methan-1-ol PubChem CID: 7537586 SMILES: OCC1=CC=CC(=C1)C1=CC=CS1