Test
2017-Fisher-Scientific-Logo
Prodotti
Applicazioni
Biologia cellulare
Applicazioni
Chimica
Genomica
Produzione e bioprocessi
Applicazioni
Produzione e bioprocessi
Proteomica
Applicazioni
Menu principale
Lab Furniture and Storage
Lab Furniture and Storage
Essential Products for Your Lab
From concept to completion, the Fisher Scientific channel offers everything you need to design, build, and fill your lab, including lab furniture, storage, workbenches, cabinets, and more. We can also help you design, plan, and install your new or remodeled lab.
Related Resources
Cell and Gene Therapy Products to Meet GMP Guidelines
Meeting good manufacturing practice guidelines for cell and gene therapy products: why your equipment matters. Discover the most common challenges faced when implementing new equipment in your manufacturing workflow and what to look for when choosing new equipment.
Cell & Gene Therapy Products to Meet GMP Guidelines
Maintain Data Integrity With Your Balance
If you want to avoid costly and reputation-damaging failed inspections, shut-downs or product recalls, maintaining data integrity is crucial. So why not choose a laboratory balance with the ALCOA principles built in?
Maintain Data Integrity with Your Balance
Six Steps to Safely Autoclave Glass Laboratory Bottles
An autoclave is a specialized piece of equipment designed to provide a physical method for disinfection and sterilization. The process is achieved using a combination of steam, high pressure, and time. Steam is remarkably effective at transferring thermal energy (compared to dry air) and can sterilise a load at 121 °C in just 15 minutes.
Six Steps to Safely Autoclave Glass Laboratory Bottles