Serological Pipets
Links
Save Now - Web Exclusive Deals
Take advantage of our web exclusive prices and place your order today.
Must Have
SUITABLE FOR TISSUE CULTURE WORK
INCREASING PIPET STABILITY IN YOUR CONTROLLER
FOR LIQUID HANDLING
Complete Your Order
Complete your serological pipet order with the necessary accessories.
Great deals
Thermo Scientific™ S1 Pipet Fillers
Ergonomically designed pipet fillers for use with graduated and volumetric glass or plastic serological pipets.
Buy NowGreat deals
Watch This Video
Corning Falcon Individually-Wrapped 100mL Serological Pipets
Featuring the highest holding capacity currently available on the market (125mL), the Corning™ Falcon™ 100mL Serological Pipet has a graduated capacity of up to 110mL. It fits easily into most media bottles and tissue culture vessels and is perfect for use in bacteriological and sterile research environments.
Buy NowGuides to Help You Choose
WHEATON Serological Pipets
Premium range, cleanroom manufactured pipets designed for your cell culture analysis. Learn about key benefits and find product order information to make your purchase today.
Thermo Scientific Nunc Serological Pipets
Highlights of the newly expanded portfolio for Nunc Serological Pipets. Find your size and packaging options, as well as ordering information to make your first purchase today.
Corning Stripette Serological Pipets
A buying guide to sterile, individually wrapped RNase-/DNase-free Stripette Serological Pipets. Triple-bagged packaging options are available that are ideal for cell-based vaccine and drug production facilities.
Falcon Tubes and Pipets
A comprehensive guide to Falcon™ Tubes, Pipets, and Pipette Controllers with key features and accessories.
Shop Your Brands
Frequently Asked Questions
What are serological pipets used for?
Serological pipets are used for transferring liquids, mixing chemical solutions or cell suspensions, and laying reagents that are composed of different densities. This is done with the help of a pipet controller, filler or bulb. Volume capacities range anywhere from <1mL up to 100mL depending on the brand and model. Serological pipets are made of plastic or glass and are commonly available sterile.
What does graduation mean?
To be able to measure the volume of liquid aspirated and dispensed, serological pipets are marked, or 'graduated', with the numerical volume on the side of the tube. Some serological pipets feature both ascending and descending graduations.
What do the abbreviations TD and TC mean?
TD stands for 'To Deliver' and TC stands for 'To Contain'. In TD-calibrated pipets, a defined amount of liquid is delivered with any residual fluid remaining on the inside walls of the tube being accounted for by previous calibration. In TC-calibrated pipets, the pipette contains a specified volume but dispenses less due to this residual liquid. As such, it is important to blow all of the liquid out of the pipet in order to have the desired volume. Graduated pipets or bulb pipets are common among TD designs, while capillary pipets are common among TC designs.