Test
Store and Protect Valuable Samples
Discover the finest selection of cold-storage solutions and supplies. We offer a comprehensive range of laboratory refrigerators and freezers, coolers and boxes, and cutting-edge cryogenic storage systems. Trust our professional-grade products to safeguard your valuable samples and optimize your storage needs. Shop now for premium cold-storage solutions.
Featured Brands
Frequently
Asked Questions
What are the different types of laboratory refrigerators?
The types of laboratory refrigerators include general-purpose refrigerators for sample and supply storage, flammable storage refrigerators for safe storage of flammable materials, explosion-proof refrigerators for hazardous environments, ultra-low temperature (ULT) freezers for long-term storage at extremely low temperatures, blood bank refrigerators for storing blood and blood products, pharmacy refrigerators for medication and vaccine storage, and cryogenic storage freezers for preserving biological samples at ultra-low temperatures.
Learn More
What is the principle of a ULT freezer?
The principle of an ultra-low temperature (ULT) freezer is based on the use of a cascade refrigeration system.
Learn More
What is the life expectancy of a ULT freezer?
The life expectancy of an ultra-low temperature (ULT) freezer can vary depending on factors such as the quality of construction, maintenance practices, and usage patterns.
Learn More
Is a laboratory freezer the same as a normal freezer?
No, a laboratory freezer is not the same as a normal household freezer. Lab freezers are specifically designed for storing temperature-sensitive materials and samples used in scientific research, healthcare, or industrial applications. They often have more precise temperature control, wider temperature ranges, and advanced features like alarms, data logging, and remote monitoring. Lab freezers may also have specialized designs, such as explosion-proof or flammable storage capabilities, to ensure safety in laboratory settings. In contrast, normal freezers are typically used for general food storage and may not provide the same level of temperature control and features required for scientific purposes.
Learn More
Science News
Lab Reporter
Webinar image
Meeting the Cold Chain Challenge
Read More
Webinar image
Finding Cold Storage Solutions
Read More
Webinar image
How to Reduce Hidden Costs in the Cell Culture Lab
Read More
Available Now
Other Programs
Sustainability Program
Sustainability Program
Read more about how we can reach net-zero goals together.
Learn More
Lab Reporter
Fisher Scientific Edge Program
Ensure quick dispatch of our most popular products.
Learn More
NLSU Program
Setting Up a New Lab?
Get all the support needed for setting up a new lab or expanding an existing lab through the NLSU program.
Learn More
NLSU Program
News Corner
Explore the latest innovations within the Fisher Scientific Channel
Learn More