Learn More
1,10-Phenanthroline monohydrate, 99+%, Thermo Scientific Chemicals
Descrizione
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1,10-Phenanthroline monohydrate is useful to study the DNA-protein interactions due to its nuclease activity. It is used as a redox indicator and reagent for spectrophotometric assay of silver. It acts as a chelating ligand for the determination of Fe, Pd and V. It also acts as a matrix metalloproteinase inhibitor. It is a photometric reagent for iron, which forms a red chelate with ferrous (iron II) ion.
Solubility
Soluble in ethanol, acetone and methanol. Slightly soluble in water and benzene.
Notes
Working solutions of 1,10-Phenanthroline monohydrate may stable for a few days at refrigerated temperatures. Incompatible with strong oxidizing agents and strong acids.

Specifica
Specifica
Materiale o nome chimico | 1,10-Phenanthroline monohydrate |
Punto di fusione | 97°C to 101°C |
Odore | Odorless |
Quantità | 250 g |
Numero UN | UN2811 |
Beilstein | 4008096 |
Indice di Merck | 14,7214 |
Sinonimo | 1,10-phenanthroline hydrate, 1,10-phenanthroline monohydrate, o-phenanthroline monohydrate, unii-ksx215x00e, 1,10-fenantrolina, 4,5-phenanthroline monohydrate, 1,10-phenanthroline, monohydrate, 1,10-fenanthrolin, 1,10-phenanthrolineo-phenanthroline, 1, 10-phenanthroline monohydrate |
Informazioni di solubilità | Soluble in ethanol,acetone and methanol. Slightly soluble in water and benzene. |
IUPAC Name | 1,10-phenanthroline;hydrate |
Vedi altri risultati |
Certificati
Certificati
Per poter ottenere i certificati è necessario indicare un numero di lotto. Per individuare il numero di lotto su ordini precedenti usi l'area dedicata allo stato degli ordini.
Numero di lotto | Tipo di certificato | Data | Catalog Number |
---|---|---|---|
10249860 | Certificato di analisi | 05/03/2024 | A14140.09,A14140.18,A14140.30 |
10247254 | Certificato di analisi | 13/07/2023 | A14140.09,A14140.18,A14140,A14140.30 |
D06Y020 | Certificato di analisi | 22/11/2021 | A14140.09,A14140.18,A14140.30 |
10112653 | Certificato di analisi | 22/11/2021 | A14140.09,A14140.18,A14140.30 |
10127179 | Certificato di analisi | 22/11/2021 | A14140.09,A14140.18,A14140.30 |
RUO – Research Use Only
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.