Learn More
Hamilton™ 1 μL 7000 Series Microliter Syringe
The 7000 series syringes are modified Microliter syringes with a plunger-in-needle design.
Marca:  Hamilton™ CAL88400
Dettagli aggiuntivi : Peso : 0.03300kg
Codice prodotto. 17804222
  • 333.00€ / Pezzo
Spedizione stimata: 21-10-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • The only zero-dead volume syringe available.
  • The fine gauge tungsten plunger wire extends to the tip of the needle, ensuring that all solvent is expelled, dispensing volumes up to 1 μL.
Specifica
Specifica
Autoclavable
22 G
Microliter Syringe
7101 KHCH
1 μL
Sharp, Bevel, Curved, Non-coring
Tungsten (Barrel)
0.145 mm
115°C
1
69.9 mm
Knurled Hub, Removable