Learn More
Keystone Cleanroom Products™ 1073B Tyvek™/HDPE Autoclave Stopper Bag Tubing
BIOPHARMAGREENER_CHOICE
Autoclave Stopper Bag Continuous Form Tubing for Steam Sterilization is made of medical grade DuPont™ Tyvek™ and HDPE film materials and yields high seal strength for stoppers, seals and heavy equipment parts. Cut tubing to desired length and heat seal both ends closed using a validateable impulse, or continuous band sealer.
Marca:  Keystone Cleanroom Products™ SBTMGTVK18
 vedi altre versioni di questo prodotto
Codice prodotto. 17718517
  • 2085.00€ / pezzi 500
Spedizione stimata: 20-01-2025
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Designed for Steam Sterilization
  • Recommended for Use in Conjunction with a Secondary Tyvek™ Sterilization Pouch
  • Hydrophobic
  • Microbial Barrier
  • Resists Tearing and Punctures
  • Lint-Free - Category (Helmke Drum)
  • Manufactured and Packaged in a ISO 6 Cleanroom
  • Certificates of Conformance sent with shipment
Specifica
Specifica
Autoclavable
Heat Seal
Single-use
Pharmaceutical Equipment
Non-sterile
<125°C
152 m
46 cm
COC
White
Steam Sterilization
Tyvek
<250°F
500 ft.
18 in.