Learn More
4-Methoxybenzyl alcohol, 98%, Thermo Scientific Chemicals
31.30€ - 446.00€
Identificatori chimici
CAS | 105-13-5 |
---|---|
Formula molecolare | C8H10O2 |
Molecular Weight (g/mol) | 138.17 |
Numero MDL | MFCD00004653 |
InChI Key | MSHFRERJPWKJFX-UHFFFAOYSA-N |
Sinonimo | 4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol |
PubChem CID | 7738 |
ChEBI | CHEBI:86918 |
IUPAC Name | (4-methoxyphenyl)methanol |
SMILES | COC1=CC=C(CO)C=C1 |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
11403017
|
Thermo Scientific Alfa Aesar
A15559.22 |
100 g |
31.30€
100g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11413017
|
Thermo Scientific Alfa Aesar
A15559.36 |
500 g |
104.00€
500g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
11423017
|
Thermo Scientific Alfa Aesar
A15559.0E |
2500 g |
446.00€
2500g |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | |||||
Descrizione
4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Methoxybenzyl Alcohol is used in the preparation of semiconductors, nanosheets and nanocrystals. Also is used as a reagent for various chemical organic reactions such as in the synthesis of quinolines. It is used as a fragrance and flavorant. It is used to study the photo catalytic oxidation of 4-methoxybenzyl alcohol to p-anisaldehyde.
Solubility
Soluble in water, chloroform, ethyl acetate.
Notes
Store in a cool, dry conditions in a well sealed container. Incompatible with oxidizing agent, acids, acid chlorides, acid anhydrides.
Identificatori chimici
105-13-5 | |
138.17 | |
MSHFRERJPWKJFX-UHFFFAOYSA-N | |
7738 | |
(4-methoxyphenyl)methanol |
C8H10O2 | |
MFCD00004653 | |
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol | |
CHEBI:86918 | |
COC1=CC=C(CO)C=C1 |
Specifica
4-Methoxybenzyl alcohol | |
22°C to 25°C | |
259°C | |
C8H10O2 | |
MFCD00004653 | |
14,604 | |
Soluble in water,chloroform,ethyl acetate. | |
COC1=CC=C(CO)C=C1 | |
138.17 | |
CHEBI:86918 | |
98% |
105-13-5 | |
1.11 | |
113°C (235°F) | |
1.545 | |
636654 | |
4-methoxybenzyl alcohol, 4-methoxyphenyl methanol, anise alcohol, anisyl alcohol, p-anisyl alcohol, p-methoxybenzyl alcohol, benzenemethanol, 4-methoxy, anisic alcohol, p-anisol alcohol, 4-methoxybenzenemethanol | |
MSHFRERJPWKJFX-UHFFFAOYSA-N | |
(4-methoxyphenyl)methanol | |
7738 | |
138.17 |
Sicurezza e movimentazione
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P310-P333+P313-P362-P501c
H315-H317-H318
missing translation for 'einecsNumber' : 203-273-6
missing translation for 'rtecsNumber' : DO8925000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
RUO – Research Use Only