Learn More
Tocris Bioscience™ AC 5216
Selective TSPO ligand
Marca:  Tocris Bioscience™ 5281/50
Codice prodotto. 18789806
  • 1150.00€ / 50mg
Esplora altre offerte speciali
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
AC 5216 is a ligand of translocator protein (TSPO, peripheral benzodiazepine receptor). Exhibits high affinity for TSPO in rat whole brain (Ki = 0.297 nM), and high potency against human and rat glial TSPO (IC50 values are 2.73 and 3.04 nM respectively). Displays no noticeable binding to a number of other receptors, transporters or ion channels. Also displays anxiolytic and antidepressant effects in rodent models, without inducing benzodiazepine-like adverse effects.
Specifica
Specifica
>98%
AC 5216
AC 5216
50 mg
401.46
For Research Use Only