Learn More
Tocris Bioscience™ Ac-Cys-NHMe
Building block for the lysine acylation using conjugating enzymes (LACE) technique
Marca: Tocris Bioscience™ 7936/10
162.42 EUR valido fino al 2024-12-16
Usa il codice promo "21630" per avere il tuo prezzo promozionale.
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Ac-Cys-NHMe is a non-canonical amino acid that is an essential building block for lysine acylation using conjugating enzymes (LACE) technique. LACE is a chemoenzymatic approach that uses an E2 small ubiquitin-like modifier (SUMO)-conjugating enzyme, Ubc9, to conjugate peptide thioesters containing a C-terminal ubiquitin-derived sequence (LRLRGG) to a lysine residue located in a recognition sequence (IKQE) on a protein via an isopeptide bond. Ac-Cys-NHMe can be reacted with a peptide hydrazide to form a reactive peptide thioester, which is the substrate for Ubc9. LACE enables the site-specific modification of proteins at lysine residues to allow attachment of biochemical probes such as fluorophores to proteins or to engineer proteins for site-specific delivery.Specifica
(2R)-2-(Acetylamino)-3-mercapto-N-methylpropanamide | |
10 mg | |
C6H12N2O2S |
10061-65-1 | |
Unnatural Amino Acids |