Learn More
Thermo Scientific™ Filtration Hardware, Accessories, and Spare Parts
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Marca: Thermo Scientific™ 710345-0010
Dettagli aggiuntivi : Peso : 0.50000kg
Includes: Three vacuum outlets, each with two-way valve and vent port; hose barb vacuum connector for 0.38 in. (9.5mm) I.D. tubing.
Descrizione
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.
Designed for use in a wide range of applications, Thermo Scientific™ Nalgene™ filtration hardware and accessories can be used in conjunction with our various lines of filtration devices.Specifica
Nalgene Reusable Filter Funnel with Clamp | |
Teflon | |
Filter Manifold Stopcock |