Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-Well Filter Plates with Ion Exchange Membrane
Optimized for a variety of applications requiring larger per well volume processing.
Marca: Cytiva (formerly Pall Lab) 8072
Dettagli aggiuntivi : Peso : 0.00100kg
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Mustang S (Cation Exchange) | |
Non-sterile | |
350 μL |
0.8 μm | |
10/Pk. |