Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration
Optimized for a variety of applications requiring larger per well volume processing.
Marca:  Cytiva (formerly Pall Lab) 8782
Dettagli aggiuntivi : Peso : 0.45400kg
Codice prodotto. 17184641
  • 230.00€ / pezzi 5
Spedizione stimata: 25-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Specifica
wwPTFE
Non-sterile
5/Pk.
0.65/0.2 μm
Solvent Filtration
2 mL