Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Solvent Filtration, Long Tip
Optimized for a variety of applications requiring larger per well volume processing.
Marca:  Cytiva (formerly Pall Lab) 8588
Dettagli aggiuntivi : Peso : 0.00100kg
Codice prodotto. 17134641
  • 310.00€ / Pezzo
Spedizione stimata: 11-09-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Specifica
wwPTFE
Non-sterile
350 μL
0.45 μm
10/Pk.