Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Optimized for a variety of applications requiring larger per well volume processing.
Marca:  Cytiva (formerly Pall Lab) 8036
Dettagli aggiuntivi : Peso : 0.94050kg
Codice prodotto. 17104611
  • 515.00€ / pezzi 10
Spedizione stimata: 25-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
  • Plates are constructed of chemically resistant, biologically-inert polypropylene
  • This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Specifica
Omega (Modified PES)
Non-sterile
10/Pk.
100 kDa
Polypropylene Housing
350 μL