Learn More
Cytiva (formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Ultrafiltration
Optimized for a variety of applications requiring larger per well volume processing.
Marca: Cytiva (formerly Pall Lab) 8034
Dettagli aggiuntivi : Peso : 0.85500kg
Descrizione
- Plates are constructed of chemically resistant, biologically-inert polypropylene
- This product is not approved for use in medical, clinical, surgical or other patient applications
Specifica
Omega (Modified PES) | |
Non-sterile | |
10/Pk. |
10 kDa | |
Polypropylene Housing | |
350 μL |