Learn More
SCHOTT AG Lighting and Imaging™ Adapter for Transmitted Light Base (TLS)
This Adapter fits the transmitted light stage to a microscope stand with base plate.
Marca: SCHOTT AG Lighting and Imaging™ 157630
Dettagli aggiuntivi : Peso : 5.00000kg
Descrizione
- Adapter for TLS 122150 Transmitted Light Base.
- Models available to fit microscope stands with base plates that have diameters of 90 mm, 100 mm, 120 mm and 180 mm.
Specifica
Adapter | |
90 mm | |
TLS 122150 Transmitted Light Base |
3.5 in. | |
Stereo Microscopy Illumination |