Learn More
Thermo Scientific™ Additional Activated Carbon Filter
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Marca: Thermo Scientific™ 50073773
1496.54 EUR valido fino al 2024-09-17
Usa il codice promo "21690" per avere il tuo prezzo promozionale.
Dettagli aggiuntivi : Peso : 12.32100kg
Descrizione
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
- Additional Activated Carbon Filter
- Fittings Included.
- Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Specifica
EN 12469 | |
Activated Carbon | |
Retrofit to 1.2/1.5 m Cabinets |
Filter | |
Fittings |