Learn More
Thermo Scientific™ Additional Activated Carbon Filter
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Marca: Thermo Scientific™ 50078783
Dettagli aggiuntivi : Peso : 0.01000kg
Descrizione
Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
- Additional Activated Carbon Filter
- Fittings Included.
- Additional exhaust carbon filters can be fitted on top of the BSC, but should only be used for traces of solvents being used as an adjunct to microbiological study.
Specifica
EN 12469 | |
Carbon | |
Retrofit to 0.9 m Herasafe KS/Safe 2020 Cabinets |
Filter | |
Fittings |